missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylbenzyl alcohol, 98%

$44.35 - $522.34

Chemical Identifiers

CAS 62285-58-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00128004
InChI Key JPEYJQDKTDVJSZ-UHFFFAOYSA-N
Synonym 2,6-dimethyl benzyl alcohol
PubChem CID 186748
IUPAC Name (2,6-dimethylphenyl)methanol
SMILES CC1=C(C(=CC=C1)C)CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61409MD View Documents Alfa Aesar™
H61409MD
250mg Each for $44.35
 
AAH6140903 View Documents Alfa Aesar™
H6140903
1g Each for $176.80
 
AAH6140906 View Documents Alfa Aesar™
H6140906
5g Each for $522.34
 
Specifications

Chemical Identifiers

62285-58-9
136.194
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
186748
CC1=C(C(=CC=C1)C)CO
C9H12O
MFCD00128004
2,6-dimethyl benzyl alcohol
(2,6-dimethylphenyl)methanol

Specifications

250mg
C9H12O
2,6-dimethyl benzyl alcohol
CC1=C(C(=CC=C1)C)CO
136.194
136.19
98%
62285-58-9
MFCD00128004
JPEYJQDKTDVJSZ-UHFFFAOYSA-N
(2,6-dimethylphenyl)methanol
186748
Solid
2,6-Dimethylbenzyl alcohol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures