Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylbenzoquinone 4-oxime, 99%

$120.20 - $120.20

Chemical Identifiers

Chemical Name or Material 2,6-Dimethylbenzoquinone 4-oxime
CAS 4965-29-1
Molecular Formula C8H9NO2
Formula Weight 151.17g/mol
MDL Number MFCD00082725
Beilstein 1941151
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1236706

 
alfa aesar™
L12367-06
5g Each for $120.20
Description & Specifications

Specifications

Melting Point ≈168°C
Quantity 5g
Odor Odorless
Chemical Name or Material 2,6-Dimethylbenzoquinone 4-oxime
CAS 4965-29-1
Assay 99%
Molecular Formula C8H9NO2
Formula Weight 151.17g/mol
MDL Number MFCD00082725
Beilstein 1941151
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.