missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylbenzoic acid, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC169010250

 View more versions of this product

Catalog No. AC169010250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dimethylbenzoic acid
Cream to White
25g
98.5
Glass bottle
99%
(CH3)2C6H3COOH
01, 259
HCBHQDKBSKYGCK-UHFFFAOYSA-N
2,6-dimethylbenzoic acid
12439
150.18
Authentic
114.0°C to 117.0°C
632-46-2
100.0
99%
C9H10O2
MFCD00002483
vic.-m-Xylylic acid
CC1=C(C(=CC=C1)C)C(=O)O
150.177
CHEBI:64827
Crystalline Powder

Chemical Identifiers

632-46-2
150.177
HCBHQDKBSKYGCK-UHFFFAOYSA-N
12439
C9H10O2
MFCD00002483
vic.-m-Xylylic acid
CHEBI:64827
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents