missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethylbenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC397560010

 View more versions of this product

Catalog No. AC397560010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Dimethylbenzaldehyde
Colorless to yellow
1123-56-4
100.0
Liquid or Low Melting Solid
97%
(CH3)2C6H3C(O)H
MFCD00128003
Authentic
1g
96.0
Glass bottle
≥96.0%
C9H10O
134.18

Chemical Identifiers

1123-56-4
100.0
C9H10O
134.18
2, 6-Dimethylbenzaldehyde
96.0
97%
(CH3)2C6H3C(O)H
MFCD00128003
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.