missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethylanisole, 98+%

$204.73 - $204.73

Chemical Identifiers

CAS 1004-66-6
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00008380
Beilstein 1858509
Synonym 2-Methoxy-m-xylene
Chemical Name or Material 2,6-Dimethylanisole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2036522 SDS Alfa Aesar™
B2036522
100g Each for $204.73
AAB2036514 SDS Alfa Aesar™
B2036514
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
2,6-Dimethylanisole is used as a pharmaceutical intermediate. Methoxymetacyclophanes is prepared from 2,6-dimethylanisole.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

1004-66-6
136.19
1858509
2,6-Dimethylanisole
C9H12O
MFCD00008380
2-Methoxy-m-xylene

Specifications

67°C (152°F)
100g
C9H12O
MFCD00008380
2-Methoxy-m-xylene
≥98%
181° to 182°C
1004-66-6
136.19
1858509
2,6-Dimethylanisole

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning