Learn More
2,6-Dimethylaniline, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC115940025
Specifications
2, 6-Dimethylaniline | |
91°C | |
Glass bottle | |
0.98 | |
Undesignated | |
2.5kg | |
87-62-7 | |
100.0 | |
C8H11N | |
MFCD00007747 | |
15, 10282 | |
Solubility in water: 7.5g/L (20 c). Other solubilities: very soluble in ether and ethanol,soluble in dimethyl sulfoxide,carbon tetrachlorid | |
CC1=C(C(=CC=C1)C)N | |
121.18 | |
CHEBI:28738 | |
Liquid |
0.9800g/mL | |
Authentic | |
1.5591 to 1.5611 | |
216°C | |
10°C to 12°C | |
99% | |
98.5 | |
98.5% min. (GC) | |
(CH3)2C6H3NH2 | |
12, 1107 | |
1-amino-2,6-dimethylbenzene, 2,6-dimethylbenzenamine, 2,6-dimethylphenylamine, 2,6-xylidine, 2,6-xylylamine, 2-amino-1,3-dimethylbenzene, 2-amino-1,3-xylene, 2-amino-m-xylene, benzenamine, 2,6-dimethyl, o-xylidine | |
UFFBMTHBGFGIHF-UHFFFAOYSA-N | |
2,6-dimethylaniline | |
6896 | |
121.18 | |
99% |
Chemical Identifiers
87-62-7 | |
121.18 | |
UFFBMTHBGFGIHF-UHFFFAOYSA-N | |
6896 | |
2,6-dimethylaniline |
C8H11N | |
MFCD00007747 | |
1-amino-2,6-dimethylbenzene, 2,6-dimethylbenzenamine, 2,6-dimethylphenylamine, 2,6-xylidine, 2,6-xylylamine, 2-amino-1,3-dimethylbenzene, 2-amino-1,3-xylene, 2-amino-m-xylene, benzenamine, 2,6-dimethyl, o-xylidine | |
CHEBI:28738 | |
CC1=C(C(=CC=C1)C)N |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 201-758-7
RTECSNumber : ZE9275000
TSCA : TSCA
Recommended Storage : product darkens if exposed to light and air