Learn More
2,6-Dimethyl-5-heptenal, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01777330
Description
It is used or fragrances designed to have a special fruity note tending towards melon, cucumber and pumpkin. It is mainly used perfuming agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,6-Dimethyl-5-heptenal | |
0.879 g/mL | |
63°C (145°F) | |
1.4441 | |
250 g | |
Air Sensitive | |
Soluble in alcohol,paraffin oil. Insoluble in water. | |
CC(CCC=C(C)C)C=O | |
140.226 | |
140.22 |
106-72-9 | |
116°C to 124°C (100 mmHg) | |
C9H16O | |
MFCD00006981 | |
UN1989 | |
2,6-dimethyl-5-heptenal, melonal, 5-heptenal, 2,6-dimethyl, bergamal, 2,6-dimethyl-5-hepten-1-al, 2,6-dimethyl-5-heptanal, 2,6-dimethylhept-5-en-1-al, fema no. 2389, 2,6-dimethyl-2-hepten-7-al, 2,6-dimethyl-5-heptenal natural | |
YGFGZTXGYTUXBA-UHFFFAOYSA-N | |
2,6-dimethylhept-5-enal | |
61016 | |
Liquid |
Chemical Identifiers
106-72-9 | |
140.226 | |
YGFGZTXGYTUXBA-UHFFFAOYSA-N | |
61016 | |
CC(CCC=C(C)C)C=O |
C9H16O | |
MFCD00006981 | |
2,6-dimethyl-5-heptenal, melonal, 5-heptenal, 2,6-dimethyl, bergamal, 2,6-dimethyl-5-hepten-1-al, 2,6-dimethyl-5-heptanal, 2,6-dimethylhept-5-en-1-al, fema no. 2389, 2,6-dimethyl-2-hepten-7-al, 2,6-dimethyl-5-heptenal natural | |
2,6-dimethylhept-5-enal |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P280-P370+P378q-P501c
H227
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber : 203-427-2
RTECSNumber : MJ8797000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only