Learn More
2,6-Dimethyl-4-nitrophenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1547206
Description
It was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt was used as internal standard in determination of 4-nitrophenol and 3-methyl-4-nitrophenol in human urine by liquid chromatography combined with tandem mass spectrometry. ,6-Dimethyl-4-nitrophenol was used to study the disposition requirement for binding of p-nitrophenol and sodium p-nitrophenolate like substrates with cyclohexaamylaose.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
2,6-Dimethyl-4-nitrophenol | |
∼168°C (decomposition) | |
MFCD00007339 | |
1873275 | |
Insoluble in water. | |
CC1=CC(=CC(=C1O)C)[N+](=O)[O-] | |
167.164 | |
167.17 |
2423-71-4 | |
C8H9NO3 | |
5 g | |
4-nitro-2,6-xylenol, 2,6-xylenol, 4-nitro, 4-nitro-2,6-dimethylphenol, phenol, 2,6-dimethyl-4-nitro, phenol,2,6-dimethyl-4-nitro, acmc-1cljn, 2, 4-nitro, intermediates-zcf02651, 2,6-dimethyl4-nitrophenol, phenol,6-dimethyl-4-nitro | |
FNORUNUDZNWQFF-UHFFFAOYSA-N | |
2,6-dimethyl-4-nitrophenol | |
17030 | |
98% |
Chemical Identifiers
2423-71-4 | |
167.164 | |
FNORUNUDZNWQFF-UHFFFAOYSA-N | |
2,6-dimethyl-4-nitrophenol |
C8H9NO3 | |
MFCD00007339 | |
17030 | |
CC1=CC(=CC(=C1O)C)[N+](=O)[O-] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
EINECSNumber : 219-353-9
RTECSNumber : ZE8225000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only