missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethyl-4-heptanone, >90% (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)

$77.65 - $77.65

Chemical Identifiers

CAS 108-83-8
Molecular Formula C9H18O
Formula Weight 142.24
MDL Number MFCD00008940
UN Number UN1224
Beilstein 1743163
Synonym Diisobutyl ketone; Isovalerone
Chemical Name or Material 2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL160150F SDS Alfa Aesar™
L160150F
2500mL Each for $77.65
AAL16015AP SDS Alfa Aesar™
L16015AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
2,6-Dimethyl-4-heptanone is used as a coating solvent. It is an active component of mint oil. It acts as a dispersant for organosol type resins. It is involved in the antigerminative treatment of bulbs and tubers. Further, it is used as a solvent for nitrocellulose. In addition to this, it acts as an intermediate in the preparation of inhibitors, active pharmaceutical ingredients and dyes.

Solubility
Miscible with ethanol, ether, carbon tetrachloride, chloroform, benzene and most organic liquids.

Notes
Incompatible with strong bases, strong oxidizing agents, strong acids and strong reducing agents.
Description & Specifications

Chemical Identifiers

108-83-8
142.24
UN1224
Diisobutyl ketone; Isovalerone
C9H18O
MFCD00008940
1743163
2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)

Specifications

108-83-8
−42°C
142.24
UN1224
2500mL
Unpleasant
2,6-Dimethyl-4-heptanone, (sum of 2,6-Dimethyl-4-heptanone & 4,6-Dimethyl-2-heptanone)
C9H18O
163° to 173°C
MFCD00008940
1743163
47°C (117°F)
Diisobutyl ketone; Isovalerone
>90%

Safety and Handling

GHS H Statement
H226-H335
Flammable liquid and vapor.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

RTECSNumber : MJ5775000

TSCA : TSCA