missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethyl-3-nitropyridine, TCI America™

$131.39 - $455.28

Chemical Identifiers

CAS 15513-52-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00023503
InChI Key AETHUDGJSSKZKT-UHFFFAOYSA-N
Synonym 3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, 3-nitro-2,6-lutidine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
PubChem CID 84958
IUPAC Name 2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09975G View Documents TCI America
N09975G
5g Each for $131.39
 
N099725G View Documents TCI America
N099725G
25g Each for $455.28
 
Specifications

Chemical Identifiers

15513-52-7
152.153
AETHUDGJSSKZKT-UHFFFAOYSA-N
84958
CC1=NC(=C(C=C1)[N+](=O)[O-])C
C7H8N2O2
MFCD00023503
3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, 3-nitro-2,6-lutidine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
2,6-dimethyl-3-nitropyridine

Specifications

105°C
38°C
15513-52-7
MFCD00023503
3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, 3-nitro-2,6-lutidine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])C
152.153
152.15
≥98.0% (GC,T)
Yellow
5g
C7H8N2O2
1325
AETHUDGJSSKZKT-UHFFFAOYSA-N
2,6-dimethyl-3-nitropyridine
84958
Crystal-Powder at 20°C
2,6-Dimethyl-3-nitropyridine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator