Please login to your online account to display your discounted pricing

2,6-Dimethyl-3,5-heptanedione, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408360250

 View more versions of this product

Catalog No. AC408360250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 156.22
Physical Form Liquid
Quantity 25g
Color Clear colorless to light yellow
Density 1.4586g/mL
CAS Max % 100.0
Name Note 99%
Chemical Name or Material 2, 6-Dimethyl-3, 5-heptanedione
MDL Number MFCD00015040
Linear Formula (CH3)2CHCOCH2COCH(CH3)2
CAS 18362-64-6
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4560 to 1.4600 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C9H16O2
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

Specifications

Chemical Name or Material 2, 6-Dimethyl-3, 5-heptanedione
Name Note 99%
CAS 18362-64-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H16O2
Linear Formula (CH3)2CHCOCH2COCH(CH3)2
Formula Weight 156.22
MDL Number MFCD00015040
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents