missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethyl-3,5-heptanedione, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC408360050

 View more versions of this product

Catalog No. AC408360050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Dimethyl-3, 5-heptanedione
5g
18362-64-6
100.0
Authentic
≥98.5%
Glass bottle
(CH3)2CHCOCH2COCH(CH3)2
156.22
Clear colorless to light yellow
99%
98.5
1.4586g/mL
Liquid
99%
C9H16O2
1.4560 to 1.4600 (20°C, 589nm)
MFCD00015040

Chemical Identifiers

18362-64-6
100.0
C9H16O2
156.22
2, 6-Dimethyl-3, 5-heptanedione, 99%
98.5
99%
(CH3)2CHCOCH2COCH(CH3)2
MFCD00015040
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning