missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Dimethyl-2-heptanol, 99%, Thermo Scientific™
$209.60 - $209.60
Chemical Identifiers
CAS | 13254-34-7 |
---|---|
Molecular Formula | C9H20O |
Molecular Weight (g/mol) | 144.258 |
MDL Number | MFCD00072198 |
InChI Key | HGDVHRITTGWMJK-UHFFFAOYSA-N |
Synonym | 2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728 |
PubChem CID | 83268 |
IUPAC Name | 2,6-dimethylheptan-2-ol |
SMILES | CC(C)CCCC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2569118
|
Thermo Scientific Chemicals
B2569118 |
50 g |
Each for $209.60
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13254-34-7 | |
144.258 | |
HGDVHRITTGWMJK-UHFFFAOYSA-N | |
83268 | |
CC(C)CCCC(C)(C)O |
C9H20O | |
MFCD00072198 | |
2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728 | |
2,6-dimethylheptan-2-ol |
Specifications
13254-34-7 | |
0.81 | |
63°C (145°F) | |
C9H20O | |
MFCD00072198 | |
2,6-dimethyl-2-heptanol, 2-heptanol, 2,6-dimethyl, dimetol, freesiol, lolitol, dimethyl heptanol, acmc-20ao86, dimetol dimethyl heptanol, dsstox_cid_21424, dsstox_rid_79728 | |
CC(C)CCCC(C)(C)O | |
144.258 | |
144.25 | |
2,6-Dimethyl-2-heptanol |
-10°C | |
179°C to 181°C | |
Characteristic | |
1.426 | |
50 g | |
HGDVHRITTGWMJK-UHFFFAOYSA-N | |
2,6-dimethylheptan-2-ol | |
83268 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
EINECSNumber : 236-244-1
RTECSNumber : MJ3324950
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only