missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethoxytoluene, 98%, Thermo Scientific Chemicals

$110.10 - $276.12

Chemical Identifiers

CAS 5673-07-4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2260614
View Documents Promotion Details
Thermo Scientific Chemicals
B2260614
25 g
Each for $110.10
 
AAB2260622
View Documents Promotion Details
Thermo Scientific Chemicals
B2260622
100 g
Each for $276.12
 
Description

Description

2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5673-07-4

Specifications

5673-07-4
222°C
Aromatic
MFCD00674003
2045327
Insoluble in water.
CC(=C)C[Mg]Cl
114.86
152.19
2,6-Dimethoxytoluene
37°C to 41°C
96°C (204°F)
C4H7ClMg
25 g
methallylmagnesium chloride, 2-methylallylmagnesium chloride, magnesium, chloro 2-methyl-2-propenyl, methallyl magnesium chloride, methallyl-magnesium chloride, 2-methylallyl magnesium chloride, bjvfgwybolmuem-uhfffaoysa-m, 2-methylallylmagnesium chloride solution, 2-methyl-2-propenyl magnesium chloride, 2-methyl-2-propen-1-yl magnesium chloride
BJVFGWYBOLMUEM-UHFFFAOYSA-M
magnesium;2-methanidylprop-1-ene;chloride
5175691
98%
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 227-131-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only