Please login to your online account to display your discounted pricing

2,6-Dimethoxypyridine 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethoxypyridine
Name Note 99%
CAS 6231-18-1
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C7H9NO2
Linear Formula (C5H3N)(OCH3)2
Formula Weight 139.15
MDL Number MFCD00006266
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115650100

  SDS
thermo fisher scientific
115650100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.15
Quantity 10g
EINECS Number 228-334-4
TSCA TSCA
Density 1.053g/mL
CAS Max % 100.0
Chemical Name or Material 2, 6-Dimethoxypyridine
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H9NO2
Linear Formula (C5H3N)(OCH3)2
MDL Number MFCD00006266
CAS 6231-18-1
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.