Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxypyridine, 98+%

$68.50 - $198.57

Chemical Identifiers

Chemical Name or Material 2,6-Dimethoxypyridine
CAS 6231-18-1
Molecular Formula C7H9NO2
Formula Weight 139.15
MDL Number MFCD00006266
Beilstein 1525332
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2274614

  SDS
alfa aesar™
B2274614
25g Each for $68.50

AAB2274622

  SDS
alfa aesar™
B2274622
100g Each for $198.57
Description & Specifications

Specifications

Formula Weight 139.15
Chemical Name or Material 2,6-Dimethoxypyridine
Assay ≥98%
Molecular Formula C7H9NO2
MDL Number MFCD00006266
CAS 6231-18-1
Beilstein 1525332
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Flash Point 61°C (141°F)
Boiling Point 178° to 180°C
Quantity 25g
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.