missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxypyridine-3-boronic acid pinacol ester, 95%

$109.81 - $246.82

Chemical Identifiers

CAS 214360-59-5
Molecular Formula C13H20BNO4
Formula Weight 265.12
MDL Number MFCD03788240
Synonym 2,6-Dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Chemical Name or Material 2,6-Dimethoxypyridine-3-boronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2728203 SDS Alfa Aesar™
H2728203
1g Each for $246.82
AAH27282MD SDS Alfa Aesar™
H27282MD
250mg Each for $109.81
Specifications Description & Specifications

Chemical Identifiers

214360-59-5
265.12
2,6-Dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
C13H20BNO4
MFCD03788240
2,6-Dimethoxypyridine-3-boronic acid pinacol ester

Specifications

39° to 42°C
214360-59-5
265.12
2,6-Dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
95%
1g
C13H20BNO4
MFCD03788240
2,6-Dimethoxypyridine-3-boronic acid pinacol ester

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning