missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dimethoxypyridine-3-boronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445410010

 View more versions of this product

Catalog No. AC445410010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 6-Dimethoxypyridine-3-boronic acid
108°C to 109°C
221006-70-8
C7H10BNO4
ADGHSWFUZUADDH-UHFFFAOYSA-N
(2,6-dimethoxypyridin-3-yl)boronic acid
2762707
97%
Glass Bottle
1g
96% min. (HPLC)
2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxy-3-pyridyl boronic acid, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxypyridine-5-boronic acid, 3-borono-2,6-dimethoxypyridine, boronic acid, 2,6-dimethoxy-3-pyridinyl
B(C1=C(N=C(C=C1)OC)OC)(O)O
182.97
182.97

Chemical Identifiers

221006-70-8
182.97
2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxy-3-pyridyl boronic acid, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxypyridine-5-boronic acid, 3-borono-2,6-dimethoxypyridine, boronic acid, 2,6-dimethoxy-3-pyridinyl
(2,6-dimethoxypyridin-3-yl)boronic acid
C7H10BNO4
ADGHSWFUZUADDH-UHFFFAOYSA-N
2762707
B(C1=C(N=C(C=C1)OC)OC)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protectio

GHS Signal Word: Warning

SDS Documents