Please login to your online account to display your discounted pricing

2,6-Dimethoxypyridine-3-boronic acid, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethoxypyridine-3-boronic acid
Assay Percent Range 97%
Molecular Formula C7H10BNO4
Formula Weight 182.97
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC445410010

 
thermo fisher scientific
AC445410010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445410050

 
thermo fisher scientific
AC445410050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.97
Quantity 1g
CAS 221006-70-8
Chemical Name or Material 2, 6-Dimethoxypyridine-3-boronic acid
Assay Percent Range 97%
Molecular Formula C7H10BNO4