Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxyphenoxyacetic acid, 98+%

$55.76 - $215.47

Chemical Identifiers

Chemical Name or Material 2,6-Dimethoxyphenoxyacetic acid
CAS 95110-10-4
Molecular Formula C10H12O5
Formula Weight 212.2
MDL Number MFCD06823823
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5089403

  SDS
alfa aesar™
H5089403
1g Each for $55.76

AAH5089406

  SDS
alfa aesar™
H5089406
5g Each for $215.47
Description & Specifications

Specifications

Formula Weight 212.2
CAS 95110-10-4
Molecular Formula C10H12O5
MDL Number MFCD06823823
Chemical Name or Material 2,6-Dimethoxyphenoxyacetic acid
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Melting Point 79° to 82°C
Assay ≥98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.