Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxynaphthalene, 99%

$82.88 - $203.42

Chemical Identifiers

Chemical Name or Material 2,6-Dimethoxynaphthalene
CAS 5486-55-5
Molecular Formula C12H12O2
Formula Weight 188.23
MDL Number MFCD00004063
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2836603

  SDS
alfa aesar™
H2836603
1g Each for $82.88

AAH2836606

  SDS
alfa aesar™
H2836606
5g Each for $203.42
Description & Specifications

Specifications

Formula Weight 188.23
Chemical Name or Material 2,6-Dimethoxynaphthalene
Assay 99%
Molecular Formula C12H12O2
MDL Number MFCD00004063
CAS 5486-55-5
Melting Point 152° to 154°C
Quantity 1g