missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxybenzoyl chloride, 97%

Manufacturer:  Alfa Aesar™ L1234514

 View more versions of this product

Catalog No. AAL1234514



Specifications

Specifications

2,6-Dimethoxybenzoyl chloride
63°C to 67°C
1989-53-3
MFCD00000665
881245
NDXRPDJVAUCBOH-UHFFFAOYSA-N
2,6-dimethoxybenzoyl chloride
74810
97%
Acrid
25g
C9H9ClO3
UN3261
benzoyl chloride, 2,6-dimethoxy, 2,6-dimethoxybenzoic acid chloride, 2,6-dimethoxybenzoylchloride, acmc-1bopm, 2,6dimethoxybenzoyl chloride, 2,6-dimethoxy-benzoylchlorid, 2,6-dimethoxylbenzoyl chloride, 2,6-dimethoxy benzoyl chloride, 2,6-dimethoxy-benzoyl chloride, benzoyl chloride,2,6-dimethoxy
COC1=C(C(=CC=C1)OC)C(=O)Cl
200.618
200.62

Chemical Identifiers

1989-53-3
200.618
NDXRPDJVAUCBOH-UHFFFAOYSA-N
74810
COC1=C(C(=CC=C1)OC)C(=O)Cl
C9H9ClO3
MFCD00000665
benzoyl chloride, 2,6-dimethoxy, 2,6-dimethoxybenzoic acid chloride, 2,6-dimethoxybenzoylchloride, acmc-1bopm, 2,6dimethoxybenzoyl chloride, 2,6-dimethoxy-benzoylchlorid, 2,6-dimethoxylbenzoyl chloride, 2,6-dimethoxy benzoyl chloride, 2,6-dimethoxy-benzoyl chloride, benzoyl chloride,2,6-dimethoxy
2,6-dimethoxybenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents