Please login to your online account to display your discounted pricing

2,6-Dimethoxybenzonitrile, 97%, ACROS Organics™

$72.66 - $72.66

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethoxybenzonitrile
CAS 16932-49-3
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9NO2
Linear Formula (CH3O)2C6H3CN
Formula Weight 163.18
MDL Number MFCD00001788
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC181720500

  SDS
thermo fisher scientific
AC181720500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC181720100

  SDS
thermo fisher scientific
181720100
Glass bottle 10g Each for $72.66
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 163.18
Quantity 50g
Color Light beige
Melting Point 119° to 123°C
Beilstein 10, 388
CAS Min % 96.0
CAS Max % 100.0
Linear Formula (CH3O)2C6H3CN
Chemical Name or Material 2, 6-Dimethoxybenzonitrile
Assay Percent Range 97%
Molecular Formula C9H9NO2
CAS 16932-49-3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
MDL Number MFCD00001788
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.