Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxybenzeneboronic acid, 97+%

$320.18 - $320.18

Chemical Identifiers

Chemical Name or Material 2,6-Dimethoxybenzeneboronic acid
CAS 23112-96-1
Molecular Formula C8H11BO4
Formula Weight 181.98
MDL Number MFCD01318987
Beilstein 3032352
Synonym 2,6-Dimethoxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2430514

 
alfa aesar™
B2430514
25g Each for $320.18
Description & Specifications

Specifications

Formula Weight 181.98
Chemical Name or Material 2,6-Dimethoxybenzeneboronic acid
Assay ≥97%
Molecular Formula C8H11BO4
Melting Point 106° to 108°C
MDL Number MFCD01318987
CAS 23112-96-1
Beilstein 3032352
Quantity 25g
Synonym 2,6-Dimethoxyphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.