Learn More
2,6-Dimethoxybenzaldehyde, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1220903
Description
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.
Solubility
Soluble in Methanol (almost transparency).
Notes
Air sensitive. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, air.
Specifications
2,6-Dimethoxybenzaldehyde | |
96°C to 98°C | |
C9H10O3 | |
1 g | |
Air Sensitive | |
Soluble in Methanol (almost transparency). | |
COC1=CC=CC(OC)=C1C=O | |
166.18 | |
166.18 |
3392-97-0 | |
285°C | |
MFCD00010862 | |
908138 | |
benzaldehyde, 2,6-dimethoxy, unii-54p7cg8v99, pubchem8226, acmc-1ckys, 2,6 dimethoxybenzaldehyde, 2,6-dimethoxy-benzaldehyde, ksc497m3h, 1,3-dimethoxy-2-formylbenzene, paragos 460025, timtec-bb sbb006541 | |
WXSGQHKHUYTJNB-UHFFFAOYSA-N | |
2,6-dimethoxybenzaldehyde | |
96404 | |
99% |
Chemical Identifiers
3392-97-0 | |
166.18 | |
WXSGQHKHUYTJNB-UHFFFAOYSA-N | |
96404 | |
COC1=CC=CC(OC)=C1C=O |
C9H10O3 | |
MFCD00010862 | |
benzaldehyde, 2,6-dimethoxy, unii-54p7cg8v99, pubchem8226, acmc-1ckys, 2,6 dimethoxybenzaldehyde, 2,6-dimethoxy-benzaldehyde, ksc497m3h, 1,3-dimethoxy-2-formylbenzene, paragos 460025, timtec-bb sbb006541 | |
2,6-dimethoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only