Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimethoxy-p-benzoquinone, 98%

$33.00 - $204.92

Chemical Identifiers

Chemical Name or Material 2,6-Dimethoxy-p-benzoquinone
CAS 530-55-2
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD00016368
Beilstein 2048182
Merck Index 14,3225
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1493603

  SDS
alfa aesar™
A1493603
1g Each for $33.00

AAA1493606

  SDS
alfa aesar™
A1493606
5g Each for $63.28

AAA1493614

  SDS
alfa aesar™
A1493614
25g Each for $204.92
Description & Specifications

Specifications

Formula Weight 168.15
Melting Point 252° to 257°C
Molecular Formula C8H8O4
RTECS Number DK4750000
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Merck Index 14,3225
MDL Number MFCD00016368
Beilstein 2048182
Chemical Name or Material 2,6-Dimethoxy-p-benzoquinone
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 530-55-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.