Learn More
2,6-Dimethoxy-4-methylbenzaldehyde, 97%, Thermo Scientific Chemicals
$434.63 - $434.63
Chemical Identifiers
CAS | 6937-96-8 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.203 |
MDL Number | MFCD01321089 |
InChI Key | VVBGEVFGPIUGNB-UHFFFAOYSA-N |
Synonym | 2,6-dimethoxy-4-methyl-benzaldehyde, benzaldehyde,2,6-dimethoxy-4-methyl |
PubChem CID | 232028 |
IUPAC Name | 2,6-dimethoxy-4-methylbenzaldehyde |
SMILES | CC1=CC(=C(C(=C1)OC)C=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2698209
|
Thermo Scientific Chemicals
H2698209 |
10 g |
Each for $434.63
|
|
|||||
Description
2,6-Dimethoxy-4-methylbenzaldehyde, is used as a pharmaceutical intermediate and in resin synthesis. It is also used as a synthetic organic chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Dimethoxy-4-methylbenzaldehyde, is used as a pharmaceutical intermediate and in resin synthesis. It is also used as a synthetic organic chemical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
Chemical Identifiers
6937-96-8 | |
180.203 | |
VVBGEVFGPIUGNB-UHFFFAOYSA-N | |
232028 | |
CC1=CC(=C(C(=C1)OC)C=O)OC |
C10H12O3 | |
MFCD01321089 | |
2,6-dimethoxy-4-methyl-benzaldehyde, benzaldehyde,2,6-dimethoxy-4-methyl | |
2,6-dimethoxy-4-methylbenzaldehyde |
Specifications
6937-96-8 | |
C10H12O3 | |
10 g | |
2,6-dimethoxy-4-methyl-benzaldehyde, benzaldehyde,2,6-dimethoxy-4-methyl | |
VVBGEVFGPIUGNB-UHFFFAOYSA-N | |
2,6-dimethoxy-4-methylbenzaldehyde | |
232028 | |
97% |
91°C to 94°C | |
MFCD01321089 | |
Air Sensitive | |
Soluble in water. | |
CC1=CC(=C(C(=C1)OC)C=O)OC | |
180.203 | |
180.2 | |
2,6-Dimethoxy-4-methylbenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only