Please login to your online account to display your discounted pricing

2,6-Dimethoxy-1,4-benzoquinone, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  331230050

 View more versions of this product

Catalog No. AC331230050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 168.15
Quantity 5g
Color Yellow
Merck Index 15, 3246
EINECS Number 208-484-7
TSCA TSCA
Melting Point 253° to 257°C
RTECS Number DK4750000
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 2, 6-Dimethoxy-1, 4-benzoquinone
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C8H8O4
MDL Number MFCD00016368
CAS 530-55-2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material 2, 6-Dimethoxy-1, 4-benzoquinone
Name Note 97%
CAS 530-55-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD00016368
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents