Please login to your online account to display your discounted pricing

2,6-Dimethoxy-1,4-benzoquinone, 97%, ACROS Organics™

$62.27 - $62.27

Chemical Identifiers

Chemical Name or Material 2, 6-Dimethoxy-1, 4-benzoquinone
Name Note 97%
CAS 530-55-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD00016368
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC331230250

  SDS
thermo fisher scientific
331230250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC331230050

  SDS
thermo fisher scientific
331230050
Glass bottle 5g Each for $62.27
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 168.15
Physical Form Crystalline Powder
Quantity 25g
Color Yellow
TSCA TSCA
EINECS Number 208-484-7
Melting Point 253° to 257°C
RTECS Number DK4750000
Merck Index 15, 3246
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Molecular Formula C8H8O4
CAS 530-55-2
Chemical Name or Material 2, 6-Dimethoxy-1, 4-benzoquinone
MDL Number MFCD00016368
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Suspected of causing cancer.
Causes skin irritation.
May cause respiratory irritation.