Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dimercaptopurine, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Dimercaptopurine
CAS 5437-25-2
Formula Weight 184.24
MDL Number MFCD00022644
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5637303

 
alfa aesar™
H5637303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 184.24
Chemical Name or Material 2,6-Dimercaptopurine
Physical Form Powder
Assay 97%
Color Pale yellow
MDL Number MFCD00022644
CAS 5437-25-2
Quantity 1g
RTECS Number UO8475000