missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Diisopropylphenyl isocyanate, 94%

Manufacturer:  Alfa Aesar™ L1061314

 View more versions of this product

Catalog No. AAL1061314



Specifications

Specifications

2,6-Diisopropylphenyl isocyanate
116°C to 117°C (10mmHg)
28178-42-9
MFCD00008882
2645721
FEUFNKALUGDEMQ-UHFFFAOYSA-N
2-isocyanato-1,3-di(propan-2-yl)benzene
92259
94%
107°C (224°F)
25g
C13H17NO
UN2206
2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato
CC(C)C1=C(C(=CC=C1)C(C)C)N=C=O
203.285
203.29

Chemical Identifiers

28178-42-9
203.285
FEUFNKALUGDEMQ-UHFFFAOYSA-N
92259
CC(C)C1=C(C(=CC=C1)C(C)C)N=C=O
C13H17NO
MFCD00008882
2,6-diisopropylphenyl isocyanate, 2,6-diisopropylphenylisocyanate, 2-isocyanato-1,3-diisopropylbenzene, benzene, 2-isocyanato-1,3-bis 1-methylethyl, isocyanic acid 2,6-diisopropylphenyl ester, isocyanic acid, 2,6-diisopropylphenyl ester, 1,3-bis 1-methylethyl-2-isocyanatobenzene, 2,6-bis 1-methylethyl phenyl isocyanate, 2-isocyanato-1,3-di propan-2-yl benzene, benzene, 1,3-bis 1-methylethyl-2-isocyanato
2-isocyanato-1,3-di(propan-2-yl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H318-H302-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

RTECSNumber : CY8545000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents