missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Diisopropylphenol, 97%, Thermo Scientific Chemicals

An anesthetic that exhibits direct activation of the GABA-A receptor

Supplier:  Thermo Scientific Chemicals L0684136

 View more versions of this product

Catalog No. AAL0684136



Description

Description

2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diisopropylphenol plays the role of an antioxidant by reacting with free radicals to form a phenoxyl radical, a property associated with all phenol-based free radical scavengers. It is an effective intravenous hypnotic agent, widely used for the induction and maintenance of anesthesia and for sedation in the intensive care unit. It is useful for the induction and maintenance of general anaesthesia.

Solubility
Very slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2,6-Diisopropylphenol
17°C to 18°C
254°C to 256°C
1.5140
500 g
14,7834
OLBCVFGFOZPWHH-UHFFFAOYSA-N
2,6-di(propan-2-yl)phenol
4943
178.28
2078-54-8
0.962g/mL
C12H18O
MFCD00008885
1866484
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
CC(C)C1=C(C(=CC=C1)C(C)C)O
178.275
CHEBI:44915
97%

Chemical Identifiers

2078-54-8
178.275
OLBCVFGFOZPWHH-UHFFFAOYSA-N
4943
2,6-di(propan-2-yl)phenol
C12H18O
MFCD00008885
propofol, 2,6-diisopropylphenol, diprivan, disoprofol, disoprivan, diisopropylphenol, ampofol, 2,6-bis 1-methylethyl phenol, fresofol, propofolum
CHEBI:44915
CC(C)C1=C(C(=CC=C1)C(C)C)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : SL0810000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only