Please login to your online account to display your discounted pricing

2,6-Diisopropylnaphthalene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Diisopropylnaphthalene
Name Note 99%
CAS 24157-81-1
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H20
Formula Weight 212.33g/mol
MDL Number MFCD00021648
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294400050

  SDS
fisher scientific acros
294400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 67° to 70°C
Color Beige
Quantity 5g
Chemical Name or Material 2, 6-Diisopropylnaphthalene
Name Note 99%
CAS 24157-81-1
CAS Min % 98.5
CAS Max % 100.0
Physical Form Beige crystalline platelets
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C16H20
Formula Weight 212.33g/mol
MDL Number MFCD00021648
EINECS Number 246-045-1
TSCA TSCA