missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',6-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2722906

 View more versions of this product

Catalog No. AAH2722906



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2',6-Dihydroxyflavone
C15H10O4
5 g
YCGXYGWBHFKQHY-UHFFFAOYSA-N
6-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
688661
97%
92439-20-8
MFCD00017676
6,2'-dihydroxyflavone, 2',6-dihydroxyflavone, 6-hydroxy-2-2-hydroxyphenyl chromen-4-one, 6-hydroxy-2-2-hydroxyphenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-2-hydroxyphenyl
C1=CC=C(C(=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O)O
254.241
254.24

Chemical Identifiers

92439-20-8
254.241
YCGXYGWBHFKQHY-UHFFFAOYSA-N
688661
C1=CC=C(C(=C1)C2=CC(=O)C3=C(O2)C=CC(=C3)O)O
C15H10O4
MFCD00017676
6,2'-dihydroxyflavone, 2',6-dihydroxyflavone, 6-hydroxy-2-2-hydroxyphenyl chromen-4-one, 6-hydroxy-2-2-hydroxyphenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 6-hydroxy-2-2-hydroxyphenyl
6-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only