Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dihydroxybenzamide, 97%

$94.93 - $349.57

Chemical Identifiers

Chemical Name or Material 2,6-Dihydroxybenzamide
CAS 3147-50-0
Molecular Formula C7H7NO3
Formula Weight 153.14
MDL Number MFCD00017125
Beilstein 2937308
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0842403

  SDS
alfa aesar™
L0842403
1g Each for $94.93

AAL0842406

  SDS
alfa aesar™
L0842406
5g Each for $349.57
Description & Specifications

Specifications

Formula Weight 153.14
CAS 3147-50-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00017125
Health Hazard 1 Warning
Chemical Name or Material 2,6-Dihydroxybenzamide
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Beilstein 2937308
Quantity 1g
Molecular Formula C7H7NO3
Melting Point ≈210°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.