Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dihydroxyacetophenone, Ultrapure MALDI Matrix

For use as a matrix in MALDI mass spectrometry analysis

Chemical Identifiers

Chemical Name or Material 2,6-Dihydroxyacetophenone
CAS 699-83-2
Name Note Ultrapure MALDI Matrix
Formula Weight 152.15
MDL Number MFCD00002270
Beilstein 1366061
Synonym DHAP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67625EXK

 
alfa aesar™
J67625EXK
5 x 10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.15
Melting Point 156° to 159°C
Chemical Name or Material 2,6-Dihydroxyacetophenone
MDL Number MFCD00002270
Synonym DHAP
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note Ultrapure MALDI Matrix
Physical Form Powder
Beilstein 1366061
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 699-83-2
Color Yellow to pale brown
Quantity 5 x 10mg
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.