missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',6'-Dihydroxyacetophenone, 99%, Thermo Scientific™

Manufacturer:  Fisher Scientific Acros 114820100

 View more versions of this product

Catalog No. AC114820100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2', 6'-Dihydroxyacetophenone
Glass bottle
156.0°C to 159.0°C
99%
98.5
99%
(HO)2C6H3COCH3
1-2,6-dihydroxyphenyl ethan-1-one, 1-2,6-dihydroxyphenyl ethanone, 2',6'-dihydroxyacetophenone, 2,6-dihydroxy acetophenone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, ethanone, 1-2,6-dihydroxyphenyl, gamma-resacetophenone, resorcinol, 2-acetyl, unii-88bo51g3y2
YPTJKHVBDCRKNF-UHFFFAOYSA-N
1-(2,6-dihydroxyphenyl)ethanone
69687
Powder
Authentic
Beige-Yellow
10g
699-83-2
100.0
C8H8O3
MFCD00002270
Solubility in water: sparingly soluble. Other solubilities: soluble in ethanol,acetone,pyridine and dmf
CC(=O)C1=C(C=CC=C1O)O
152.15
152.15
99%

Chemical Identifiers

699-83-2
152.15
YPTJKHVBDCRKNF-UHFFFAOYSA-N
69687
CC(=O)C1=C(C=CC=C1O)O
C8H8O3
MFCD00002270
1-2,6-dihydroxyphenyl ethan-1-one, 1-2,6-dihydroxyphenyl ethanone, 2',6'-dihydroxyacetophenone, 2,6-dihydroxy acetophenone, 2,6-dihydroxyacetophenone, 2-acetylresorcinol, ethanone, 1-2,6-dihydroxyphenyl, gamma-resacetophenone, resorcinol, 2-acetyl, unii-88bo51g3y2
1-(2,6-dihydroxyphenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 211-833-6

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only