Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Dihydroxy-4-methylpyridine, 98+%

$213.96 - $687.08

Chemical Identifiers

Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine
CAS 4664-16-8
Molecular Formula C6H7NO2
Formula Weight 125.13
MDL Number MFCD00040464
Synonym 2,6-Dihydroxy-4-picoline; 6-Hydroxy-4-methyl-2-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2786003

  SDS
alfa aesar™
H2786003
1g Each for $213.96

AAH2786006

  SDS
alfa aesar™
H2786006
5g Each for $687.08
Description & Specifications

Specifications

Formula Weight 125.13
Quantity 1g
MDL Number MFCD00040464
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H7NO2
CAS 4664-16-8
Synonym 2,6-Dihydroxy-4-picoline; 6-Hydroxy-4-methyl-2-pyridone
Health Hazard 1 Warning
Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 195° to 198°C
Assay ≥98%

Applications
In order to synthesize macrocycles containing pyridine, 2,6-his-(bromomethyl)pyridine was prepared by bromonating 2,6-di (hydroxymethyl)pyridine.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.