Please login to your online account to display your discounted pricing

2,6-Difluorotoluene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorotoluene
Name Note 99%
CAS 443-84-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6F2
Linear Formula F2C6H3CH3
Formula Weight 128.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC320010050

  SDS
thermo fisher scientific
320010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC320010010

  SDS
thermo fisher scientific
AC320010010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 128.12
Quantity 5g
Color Clear colorless
TSCA TSCA
Density 1.1200g/mL
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
CAS Min % 98.5
Refractive Index 1.4515 to 1.4535 (20°C, 589nm)
Name Note 99%
Linear Formula F2C6H3CH3
Molecular Formula C7H6F2
MDL Number MFCD00043898
CAS 443-84-5
Assay Percent Range 99%
Chemical Name or Material 2, 6-Difluorotoluene
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.