Please login to your online account to display your discounted pricing

2,6-Difluorotoluene, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC320010010

 View more versions of this product

Catalog No. AC320010010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 128.12
Quantity 1g
Color Clear colorless
TSCA TSCA
Density 1.1200g/mL
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Refractive Index 1.4515 to 1.4535 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00043898
Chemical Name or Material 2, 6-Difluorotoluene
Name Note 99%
Linear Formula F2C6H3CH3
Assay Percent Range 99%
CAS 443-84-5
Molecular Formula C7H6F2

Specifications

Chemical Name or Material 2, 6-Difluorotoluene
Name Note 99%
CAS 443-84-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H6F2
Linear Formula F2C6H3CH3
Formula Weight 128.12
MDL Number MFCD00043898
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents