Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorotoluene, 98%

Chemical Identifiers

Chemical Name or Material 2,6-Difluorotoluene
CAS 443-84-5
Molecular Formula C7H6F2
Formula Weight 128.12
MDL Number MFCD00043898
UN Number UN2388
Beilstein 1932656
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2507603

 
alfa aesar™
B2507603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2507614

 
alfa aesar™
B2507614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2507606

 
alfa aesar™
B2507606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.12
Chemical Name or Material 2,6-Difluorotoluene
Assay 98%
Molecular Formula C7H6F2
MDL Number MFCD00043898
CAS 443-84-5
UN Number UN2388
Beilstein 1932656
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES
Flash Point 10°C (50°F)
Odor Aromatic
Boiling Point 112° to 115°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.