Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluoropyridine-3-carboxaldehyde, 95%

Chemical Identifiers

Chemical Name or Material 2,6-Difluoropyridine-3-carboxaldehyde
CAS 155601-65-3
Formula Weight 143.09
MDL Number MFCD08277633
Beilstein 7757317
Synonym 2,6-Difluoronicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6442203

 
alfa aesar™
H6442203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6442206

 
alfa aesar™
H6442206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.09
Boiling Point 64° to 66°C (15mmHg)
Melting Point 19° to 20°C
MDL Number MFCD08277633
Density 1.378g/mL
Synonym 2,6-Difluoronicotinaldehyde
Refractive Index 1.4920
Beilstein 7757317
Chemical Name or Material 2,6-Difluoropyridine-3-carboxaldehyde
Assay 95%
Storage Note 1 -30° to -10°C
Physical Form Liquid
Sensitivity Moisture Sensitive
Quantity 1g
CAS 155601-65-3
Safety and Handling

-30° to -10°C