Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',6'-Difluoropropiophenone, 97%

$38.18 - $142.07

Chemical Identifiers

Chemical Name or Material 2',6'-Difluoropropiophenone
CAS 85068-31-1
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00015506
Beilstein 7368898
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2009703

  SDS
alfa aesar™
B2009703
1g Each for $38.18

AAB2009706

  SDS
alfa aesar™
B2009706
5g Each for $142.07
Description & Specifications

Specifications

Formula Weight 170.16
Chemical Name or Material 2',6'-Difluoropropiophenone
Assay 97%
Molecular Formula C9H8F2O
MDL Number MFCD00015506
CAS 85068-31-1
Beilstein 7368898
Boiling Point 70° to 71°C (4mmHg)
Quantity 1g