Please login to your online account to display your discounted pricing

2,6-Difluorophenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorophenylboronic acid
CAS 162101-25-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.5%
Assay Percent Range 97%
Molecular Formula C6H5BF2O2
Formula Weight 157.91
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC359150010

  SDS
fisher scientific acros
35915-0010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC359150050

  SDS
thermo fisher scientific
35915-0050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.5%
Physical Form Crystalline Powder
Formula Weight 157.91
Quantity 1g
Color White to off-white
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 6-Difluorophenylboronic acid
Assay Percent Range 97%
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Molecular Formula C6H5BF2O2
CAS 162101-25-9
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.