Please login to your online account to display your discounted pricing

2,6-Difluorophenylacetonitrile, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorophenylacetonitrile
CAS 654-01-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H5F2N
Linear Formula F2C6H3CH2CN
Formula Weight 153.13
MDL Number MFCD00009973
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311670010

  SDS
thermo fisher scientific
311670010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 153.13
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.2300g/mL
CAS Max % 100.0
Linear Formula F2C6H3CH2CN
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2, 6-Difluorophenylacetonitrile
Refractive Index 1.4800 to 1.4820 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C8H5F2N
CAS 654-01-3
MDL Number MFCD00009973
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.