Please login to your online account to display your discounted pricing

2,6-difluorophenylacetone, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 6-Difluorophenylacetone
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H8F2O
Linear Formula (F)2C6H3CH2COCH3
Formula Weight 170.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC379150050

  SDS
fisher scientific acros
AC379150050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379150250

  SDS
thermo fisher scientific
AC379150250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 170.16
Quantity 5g
Color Clear colorless
CAS 101712-20-3
Chemical Name or Material 2, 6-Difluorophenylacetone
Infrared Spectrum Authentic
Molecular Formula C9H8F2O
Linear Formula (F)2C6H3CH2COCH3
Refractive Index 1.4790 to 1.4820 (20°C, 589nm)
Assay Percent Range 98%