Please login to your online account to display your discounted pricing

2,6-difluorophenylacetone, ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC379150250

 View more versions of this product

Catalog No. AC379150250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5% (GC)
Formula Weight 170.16
Quantity 25g
Color Clear colorless
Chemical Name or Material 2, 6-Difluorophenylacetone
Assay Percent Range 98%
Molecular Formula C9H8F2O
CAS 101712-20-3
Infrared Spectrum Authentic
Linear Formula (F)2C6H3CH2COCH3
Refractive Index 1.4790 to 1.4820 (20°C, 589nm)

Specifications

Chemical Name or Material 2, 6-Difluorophenylacetone
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C9H8F2O
Linear Formula (F)2C6H3CH2COCH3
Formula Weight 170.16