missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Difluorophenylacetic acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC296630050

 View more versions of this product

Catalog No. AC296630050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 6-Difluorophenylacetic acid
98° to 101°C
Authentic
85068-28-6
100.0
Crystalline Powder
99%
F2C6H3CH2COOH
MFCD00010001
White
5g
99%
98.5
Glass bottle
≥98.5%
C8H6F2O2
172.13

Chemical Identifiers

85068-28-6
100.0
C8H6F2O2
172.13
2, 6-Difluorophenylacetic acid, 99%
98.5
99%
F2C6H3CH2COOH
MFCD00010001
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 285-289-3

TSCA : TSCA