Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorophenyl isothiocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2,6-Difluorophenyl isothiocyanate
CAS 207974-17-2
Molecular Formula C7H3F2NS
Formula Weight 171.17
MDL Number MFCD00041047
UN Number UN2922
Beilstein 2090586
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1190303

  SDS
alfa aesar™
L1190303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1190306

  SDS
alfa aesar™
L1190306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 171.17
Chemical Name or Material 2,6-Difluorophenyl isothiocyanate
Assay 97%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C7H3F2NS
MDL Number MFCD00041047
CAS 207974-17-2
UN Number UN2922
Beilstein 2090586
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Flash Point 104°C (219°F)
Boiling Point 217°C
Quantity 1g

Applications
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with amines, alcohols and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.