missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,6-Difluorophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1190303

 View more versions of this product

Catalog No. AAL1190303



Description

Description

Applications
2,6-Difluorophenyl isothiocyanate is used as chemical, organic and pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture. Incompatible with amines, alcohols and oxidizing agents.
Specifications

Specifications

2,6-Difluorophenyl isothiocyanate
217°C
207974-17-2
MFCD00041047
2090586
DBSXNGIBAKYMSS-UHFFFAOYSA-N
1,3-difluoro-2-isothiocyanatobenzene
4031601
97%
104°C (219°F)
1g
C7H3F2NS
UN2922
2,6-difluorophenyl isothiocyanate, 2,6-difluorophenylisothiocyanate, benzene, 1,3-difluoro-2-isothiocyanato, 2,6-difluorobenzenisothiocyanate, pubchem5021, acmc-1caus, 2,6-difluorophenyl-isothiocyanate, 1,3-difluoro-2-isothiocyanato-benzene, benzene,1,3-difluoro-2-isothiocyanato, 2,6-difluorophenyl isothiocyanate
C1=CC(=C(C(=C1)F)N=C=S)F
171.165
171.17

Chemical Identifiers

207974-17-2
171.165
DBSXNGIBAKYMSS-UHFFFAOYSA-N
4031601
C1=CC(=C(C(=C1)F)N=C=S)F
C7H3F2NS
MFCD00041047
2,6-difluorophenyl isothiocyanate, 2,6-difluorophenylisothiocyanate, benzene, 1,3-difluoro-2-isothiocyanato, 2,6-difluorobenzenisothiocyanate, pubchem5021, acmc-1caus, 2,6-difluorophenyl-isothiocyanate, 1,3-difluoro-2-isothiocyanato-benzene, benzene,1,3-difluoro-2-isothiocyanato, 2,6-difluorophenyl isothiocyanate
1,3-difluoro-2-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents